Digi-Sense Food Infrared Thermometer
Ideal for food inspections and HACCP programs
  • Non-contact measurement eliminates the risk of cross-contamination
  • Scan numerous surfaces quickly and easily
  • Sealed, hand-washable IP54 housing
Temperature measurement is essential when ensuring proper food storage, cooking and serving environments, and is required to be FDA compliant. Quick checks with a non-contact thermometer can remove the hazard of cross-contamination and reduces cycle time over a traditional penetration probe. This thermometer provides this safe, non-contact reading in under a second.
Use the TempTestr ® to monitor HACCP danger zone temperature standards for food (40 to 140°F or 4 to 60°C), the critical range where harmful bacteria grows most rapidly. LED’s on the meter quickly indicate if the food temperature is within a green or red bacterial growth zone. A green LED light indicates food-safe hot and cold holding temperatures. Red LED light indicates that food is exposed to a potentially dangerous zone.
ITEM(S) ON SALE
$107.96 - $333.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Max Temperature (° F) Description
Availability Pricing
COMPARE
2.5:1 400 Food Infrared (IR) Thermometer
$107.96USD / Each
$119.96USD / Each
COMPARE
2.5:1 400 Food Infrared (IR) Thermometer with NIST-Traceable Calibration
$333.00USD / Each