Cole-Parmer® Irreversible High-Temperature 1-Point Square Labels / Indicators
Reliable proof that the product was maintained at the correct temperature
  • Used as a quick inspection tool that the product is in usable condition during the shipping and receiving process
  • Save time and money due to costly recalls
  • Self-adhesive design
Labels are made up of a series of temperature-sensitive elements sealed between heat-resistant substrates with transparent windows. Each element changes color distinctly as its rated temperature is met or exceeded. The changes to the label are irreversible, providing a temperature history of the surface being monitored. The labels will not delaminate when removed, allowing them to be attached to an inspection report to serve as a permanent record. Ideal for use in the food, automotive, molding and metal working industries. Accuracy is ±1.8°F below 212°F and ±1°F above 212°F. Shelf life: 1 year after date of purchase.
$26.40 - $41.20USD / Pkg of 50
30 variations of this product are available.
See All Product Options Below
30 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Height (in) Number of Temperature Indicators Temperature Points (° F)
Availability Pricing
COMPARE
1/2 1 84
$33.60USD / Pkg of 50
COMPARE
1/2 1 93
$41.20USD / Pkg of 50
COMPARE
1/2 1 99
$33.60USD / Pkg of 50
COMPARE
1/2 1 104
$33.60USD / Pkg of 50
COMPARE
1/2 1 108
$33.60USD / Pkg of 50
COMPARE
1/2 1 111
$33.60USD / Pkg of 50
COMPARE
1/2 1 120
$26.40USD / Pkg of 50
COMPARE
1/2 1 129
$30.10USD / Pkg of 50
COMPARE
1/2 1 149
$26.40USD / Pkg of 50
COMPARE
1/2 1 199
$26.40USD / Pkg of 50
COMPARE
1/2 1 241
$26.40USD / Pkg of 50
COMPARE
1/2 1 280
$31.20USD / Pkg of 50
COMPARE
1/2 1 300
$31.20USD / Pkg of 50
COMPARE
1/2 1 340
$26.40USD / Pkg of 50
COMPARE
1/2 1 360
$26.40USD / Pkg of 50
COMPARE
1/2 1 399
$30.10USD / Pkg of 50
COMPARE
1/2 1 450
$26.40USD / Pkg of 50
COMPARE
1/2 1 480
$26.40USD / Pkg of 50
COMPARE
1/2 1 554
$26.40USD / Pkg of 50
COMPARE
1/2 1 115
$41.20USD / Pkg of 50
Showing 1 - 20 of 30