Digi-Sense™ Precalibrated Compact AC and AC/DC Clamp Meters
Compact design for testing in tough-to-reach places
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards
  • Increased durability with double-molded plastic housing
  • Instant visibility with the LCD alert when measuring noncontact AC voltage
  • Access difficult testing areas with the 1-3/16" (30 mm) jaw opening
  • Improved measurement ease with data Hold function on the large backlit display
These smaller-sized autoranging clamp meters feature diode testing, continuity check with a buzzer sound at 50 Ω, and a measurement rate of 2 readings per second. Additional functions include data Hold, overrange and low-battery indication, and auto power-off. Model 20250-56 also measures DC current and capacitance, and features a peak Hold function that captures the peak AC current or voltage reading.
ITEM(S) ON SALE
$40.00 - $64.80USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (amps) Current (AC/DC)
Availability Pricing
COMPARE
400 No
$40.00USD / Each
$50.00USD / Each
COMPARE
400 Yes
$64.80USD / Each
$81.00USD / Each