Traceable® Precalibrated Thermohygrometers
Ideal for use in the HVAC, IAQ, and environmental industries
  • Reliable measurements, guaranteed! Precalibrated to NIST-traceable standards.
  • ±2% basic RH accuracy
The Traceable handheld thermohygrometers provide accurate and reliable readings in an ergonomic design. Ideal for use in HVAC, IAQ, laboratories, cleanrooms, warehouses, factories and storage areas. Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
Large backlit LCD displays humidity, dew point, wet bulb, and temperature readings. Other features include Min/Max and Hold functions, user-selectable temperature readout in °F or °C, integrated humidity and temperature (NTC thermistor) sensor, and auto power-off.
$196.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
COMPARE
3 AAA (included) 100 212
$196.00USD / Each