Cole-Parmer® Quick Disconnect Thermocouple Probe Handles (Only)
Designed to install detachable probes directly into the handle
  • Handle adaptor excepts detachable probes come with a mini connectors
  • Compatible with several different designs of detachable probes.
  • Ideal for switching out probes when working with multiple applications
  • Handle includes a 5ft coil cord with mini connector
  • Each probe comes with a color coded ANSI coiled cable: type J-black, type K-yellow, type T-blue, and type E-purple
  • Custom designs or changes to this product are available upon request
Note: handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
$49.50 - $51.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
200 K
$49.50USD / Each
COMPARE
200 J
$51.50USD / Each