Digi-Sense Red Spirit Refrigerator Glass Thermometers, Celsius-scale
Replace mercury thermometers for a specific application with the same accuracy without any of the hazards
  • Non-mercury liquid
These thermometers are designed for use in specific applications. These Durac thermometers feature markings that are permanently printed on the glass, individually serialized, and red spirit liquid that has a dark scale contrasts against a white background. Some thermometers are PTFE coated for added safety which may cause a slight change in accuracy up to 1 division.
These Refrigerator thermometers - indicate temperature ranges at 4°C and 6°C marked with arrows.
$47.00 - $71.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
COMPARE
20 -5 152
$47.00USD / Each
COMPARE
20 -5 152
$71.50USD / Each