Digi-Sense® Temp Series Thermocouple Thermometers
Heavy-duty thermometer offers versatility and accuracy in a compact design
  • 3-way hands-free operation available
  • Large easy-to-read screen with back light
  • Rugged armor with built-in stand—withstand the toughest conditions
  • Easy-to-use automatic field calibration ensures accurate readings
  • Probes are sold separately
These robust and reliable meters work in any environment from the lab to the production floor. The sturdy armored construction protects against knocks and jars. All models are ergonomically designed for both large and small hands alike. The sealed keypad and ABS plastic case meet IP54 standards for splash resistance. Select from the three most popular thermocouple types to meet all your basic temperature measurement needs.
Digi-Sense's exclusive “3-WAY HANDS-FREE” capability allows “hands-free” use—use the built-in stand on a table top, attach the meter to metal objects through magnets inserted into the armor boot, or hang from a pipe or belt using the hook-and-loop strap. Meters come standard with the integral stand; the “hands-free” option adds magnets and strap (order separately).
ITEM(S) ON SALE
$318.75 - $497.25USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Types Of Probes Accepted Description
Availability Pricing
COMPARE
1200 Type J Temp10 Single-Input Type J Thermocouple Thermometer
$318.75USD / Each
$375.00USD / Each
COMPARE
1372 Type K Temp10 Single-Input Type K Thermocouple Thermometer
$318.75USD / Each
$375.00USD / Each
COMPARE
1372 Type T Temp10 Single-Input Type T Thermocouple Thermometer
$318.75USD / Each
$375.00USD / Each
COMPARE
1800 Type E, Type J, Type K, Type T Temp100 Dual-Input Thermocouple Thermometer
$433.50USD / Each
$510.00USD / Each
COMPARE
1768 Type B, Type E, Type J, Type K, Type N, Type R, Type S, Type T Temp-300 Dual-Input Datalogging Thermocouple Thermometer with Boot
$497.25USD / Each
$585.00USD / Each