Digi-Sense Thermocouple Industrial Probes Assembly - MgO Insulated Type J
These MgO thermocouple assemblies consist of a single or dual thermocouple element constructed of various sheath materials, assembled together with a connection head.
FEATURES
  • Inconel, 316 Stainless steel or 304 Stainless steel sheath materials
  • 1/2'' X 1/2'' NPT stainless steel hex nipple for head mounting fittings
  • Aluminum screw cover, Cast iron screw cover, or Cast iron/aluminum explosion proof class C Heads
  • Standard Limits of Error
  • Grounded Measuring Junction
  • Custom built assemblies available upon request
$152.00 - $158.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Sheath material
Availability Pricing
COMPARE
$152.00USD / Each
COMPARE
$158.00USD / Each
COMPARE
$155.00USD / Each
COMPARE
$158.50USD / Each