Digi-Sense Thermocouple Industrial Probes Assembly, MgO Insulated Type K
Davis Instruments can provide many additional configurations beyond those listed here
  • 316 stainless steel sheath
Magnesium Oxide insulated thermocouples, commonly referred to as MgO thermocouples, are used in many process and laboratory applications. Many desirable characteristics make them an excellent choice for general and special purpose applications. They are available in all thermocouple element types, single or dual elements, a wide variety of sheath diameters / materials, rugged in nature, bendable, and have fairly high temperature ratings. The thermocouples listed are commonly used; however, Davis Inotek Instruments can provide many additional configurations beyond those listed.
$55.50 - $132.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Probe Type Sheath Length (in) Description
Availability Pricing
COMPARE
J 6 Probe Type-J 6\" L Re-Aj Fg Spld
$72.00USD / Each
COMPARE
K 10 Probe Type-K 10\" L Hi-Tmp Fgls
$55.50USD / Each
COMPARE
K 12 Probe Type-K 12\" L Fg SS Stplug
$71.00USD / Each
COMPARE
K 15 Temperature Probe, Type K, 15\" L, Heatshrink and Fiberglass
$94.50USD / Each
COMPARE
K 9 Temperature Probe, Type K, 9\" L, Fiberglass, Stainless Overbraid
$94.50USD / Each
COMPARE
K 12 Probe Type-K 12\" L Fg SS Stjack
$96.80USD / Each
COMPARE
K 24 Probe Type-K 24\" L Hi-Tmp Fb Thermocouple Probe
$132.00USD / Each
COMPARE
K 12 Temperature Probe, Type K, 12\" L, Heatshrink and Fiberglass
$91.50USD / Each
Showing 1- 8 of 8