Digi-Sense Thermocouple Uniwell - Universal Industrial Probes
A low cost alternative to thermowell assemblies
The Uniwell eliminates the need for thermowells and spring loaded probes. With a unique adjustable feature that allows the probe to be customized to the required insertion length, through the use of an adjustable ½” NPT compression fitting. Also designed to be directly inserted into the medium being measured without the use of a thermowell. This allows for ease of installation where a thermowell is not required. The probes conductor is made of 14 gauge wire and is encased with Magnesium Oxide insulation, and has a 446 stainless steel heavy wall sheath producing an increased life span. Featuring a standard size aluminum protection head with a gasketed cap to seal out dust and moisture and houses a ceramic terminal block. Head has a 3/4" NPT(F) opening on the side for easy connection to
$271.75USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
1400 J
$271.75USD / Each
COMPARE
2000 K
$271.75USD / Each