Digi-Sense Thermowells - 316 Stainless Steel
Designed for easy replacement of your industrial probe without shutting the operation down.
To match your probe to a thermowell, first measure the distance from the probe’s tip to the top of the thread (typical thread length is 0.5" for 1/2" NPT fittings). Next, match this length to the probe depth of the thermowell (“A” dimension). Other thermowell sizes and materials are available—call our Application Specialists for more information.
Limited-Space Thermowell when you need a shorter insertion depth.
Standard Thermowell for general-purpose applications.
Heavy-Duty Thermowell for higher fluid velocity ratings.
$27.90 - $204.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Diameter (in) Length (in) Material
Availability Pricing
COMPARE
316 Stainless Steel
$27.90USD / Each
COMPARE
316 Stainless Steel
$33.40USD / Each
COMPARE
316 Stainless Steel
$45.90USD / Each
COMPARE
316 Stainless Steel
$91.80USD / Each
COMPARE
316 Stainless Steel
$204.00USD / Each
COMPARE
316 Stainless Steel
$34.30USD / Each
COMPARE
316 Stainless Steel
$47.50USD / Each
COMPARE
316 Stainless Steel
$92.80USD / Each
COMPARE
316 Stainless Steel
$141.75USD / Each
Showing 1- 9 of 9