Masterflex® Digital Flowmeter Cables and Adapters
Displays instantaneous, total and accumulated flow rates.
  • Built-in Flow Linearizer (10 point linearization of the flow curve)
  • Up to 47 different volumetric and mass flow engineering units
  • Large 13mm (0.51") digits for flow rate and 5.5mm (0.21") for Total
  • Digital RS-232 interface (multi-drop capability of up to 64 devices requires RS-485)
  • Compact design for unit mount, panel mount, wall mount or field mount applications
  • User-programmable, optically-isolated pulse output
  • Two programmable, optically-isolated, digital outputs
  • Flow controllers, set point command control via local LCD or digital interface
  • Programmable set point table with ramping up/down capability up to 16 steps
  • Free configuration and monitoring utility software
Use these totalizers for flowmeters and controllers with a voltage or current input and output interface where flow indications / control and totalizers, or alarm functions are required. These flow totalizers can be used when re-transmission of the flow rate and/or totalizer functions via optically-isolated pulse output or serial communication is desired.Local or programmable set point control for flow controllers are easily done via keypad. Activation of user-supplied equipment via programmable optically-isolated digital outputs are easily done when flow alarms or totalizers events are active.
The graphical LCD display has large ½” (13 mm) digits for flow rate and 0.21" (5.5 mm) for total and can be set by you to simultaneously show different combination of the flow parameters: flow rate, totalizers, flow alarms, and diagnostic events. All configuration parameter settings are easily accessed via a simple user-interface menu driven by a 6 button key-pad which can be password-protected.
Totalizer kits, below, enable you to power your totalizer depending on your input/output power requirements, for both flowmeters and controllers, and are required if you have no power or communication cables. Kits 32650-92 and -94 are mounting brackets only, and require user-supplied custom cables.
$12.00 - $311.00USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Aalborg Totalizer for Gas Mass Flowmeters and Controllers with Input and Output
Buy More Save More
EW-32650-22 Mfr # TIO-LAA2
COMPARE
Totalizer for Gas Mass Flowmeters and Controllers with Input and Output
$311.00USD / Each
Aalborg GFM Flowmeter Mounting Kit, w/ Communication Branch
Buy More Save More
EW-32650-80 Mfr # KIT-TM-DD
COMPARE
GFM Flowmeter Mounting Kit, w/ Communication Branch
$49.00USD / Each
Aalborg Flow meter mounting kit, flat wire cable between GFM and TIO9 pin D-connectors
Buy More Save More
EW-32650-84 Mfr # KIT-TM-FD
COMPARE
Flow meter mounting kit, flat wire cable between GFM and TIO9 pin D-connectors
$22.00USD / Each
Aalborg GFC Flowmeter Mounting Kit, w/ Digital Communication Branch
Buy More Save More
EW-32650-86 Mfr # KIT-TC-110NA--2C
COMPARE
GFC Flowmeter Mounting Kit, w/ Digital Communication Branch
$79.00USD / Each
Aalborg GFC Flowmeter Mounting Kit, w/ Communication Branch
Buy More Save More
EW-32650-88 Mfr # KIT-TC-110NA--4C
COMPARE
GFC Flowmeter Mounting Kit, w/ Communication Branch
$79.00USD / Each
Aalborg GFC Flowmeter Mounting Kit, with 110 VAC to 24 VDC Power Supply
Buy More Save More
EW-32650-90 Mfr # KIT-TC-110NA--4N
COMPARE
GFC Flowmeter Mounting Kit, with 110 VAC to 24 VDC Power Supply
$74.00USD / Each
Aalborg Flow meter and totalizer mounting kit, no cables
Buy More Save More
EW-32650-92 Mfr # KIT-TM
COMPARE
Flow meter and totalizer mounting kit, no cables
$12.00USD / Each
Aalborg Flow controller and totalizer mounting kit, no cables
Buy More Save More
EW-32650-94 Mfr # KIT-TC
COMPARE
Flow controller and totalizer mounting kit, no cables
$12.00USD / Each
Aalborg Flow Totalizer Accessory, 9 Conductor Round Shielded Cable
Buy More Save More
EW-32650-96 Mfr # CBL-DA9-X
COMPARE
Flow Totalizer Accessory, 9 Conductor Round Shielded Cable
$50.00USD / Each
COMPARE
Power Cable 6-ft, 12 Pin Shielded M16
$174.20USD / Each
Aalborg CBL-XFM Power and communication cable with 15-pin D-connector
Buy More Save More
EW-33401-90 Mfr # CBL-XFM
COMPARE
Power and communication cable with 15-pin D-connector
$49.00USD / Each
Aalborg PS-XFM-110NA-2 Power Supply and Communication Cable, 115 VAC to 15-Pin D-Connector
Buy More Save More
EW-33401-91 Mfr # PS-XFM-110NA-2
COMPARE
Power Supply and Communication Cable, 115 VAC to 15-Pin D-Connector
$64.00USD / Each
Aalborg Power supply with communication and 9-wire analog output
Buy More Save More
EW-33401-92 Mfr # PS-XFM-110NA-2-A
COMPARE
Power supply with communication and 9-wire analog output
$102.00USD / Each
Showing 1- 13 of 13