Aalborg® Digital Gas Mass Flowmeter
Stores calibration data for up to 10 gases
  • Self-diagnostic tests
  • Automatic sensor zero offset adjustment
These digital flowmeters represent a new generation in mass flow technology. Meters incorporate a precision microcontroller and nonvolatile memory that stores all hardware specific variables, up to 10 different calibration tables, conversion factors for up to 32 gases, and supports up to 23 volumetric flow or mass flow engineering units, including user defined units. This feature allows the same meter to be calibrated for multiple gases while maintaining the rated accuracy on each. In addition, provision is made for a user-defined conversion factor. Conversion factors may be applied to any of the ten gas calibrations via digital interface commands. Program flowmeters remotely via RS-485 or RS-232. Each unit comes with a 15-pin D-connector with a stripped 6-ft, 3-wire computer communications cable and a 3-ft, 2-wire cable for connection to a power supply. Additional cables as well as power supplies for the flowmeters are available below.
Flowmeters support various functions including programmable flow totalizer, high and low flow alarm, automatic zero adjustment, two relay outputs, jumper selectable 0 to 5 VDC or 4 to 20 mA analog outputs, status LED diagnostic, and internal or user-specific K-factors. LCD display with adjustable back light provides flow, total, and diagnostic reading simultaneously. The digital RS485 and RS232 interfaces provides access to applicable internal data including flow, CPU temperature, auto zero, totalizer and alarms settings, gas table, conversion factors and engineering units selection, dynamic response compensation and linearization table adjustment. The analog interface provides 0 to 5 VDC or 4 to 20 mA outputs for flow reading. The auto zero feature necessitates a condition of absolutely no flow through the meter during the adjustment process. Provisions are made to either start, read, or save the current auto zero value via digital commands.
The total volume of the gas is calculated by integrating the actual gas flow rate as a function of time. Program high and low gas flow alarm limits via digital interface. Alarm action can be assigned with preset delay interval (0 to 3600 seconds) to activate the contact closure (separate for high and low alarm). Latch mode control feature allows each relay to be latched on or follow the corresponding alarm status.
Lower-cost units are also available without LCD for OEM applications in cooling systems, semiconductor manufacturing, or gas chromatography applications. Meters with Profibus industrial communication capabilities are also available. Contact an Applications Specialist for additional information regarding these options.
Please note that these mass flowmeters are designed to work only with clean gases. Never try to measure flow rates of liquids with these meters.
$852.00 - $1,604.00USD / Each
149 variations of this product are available.
See All Product Options Below
149 PRODUCT OPTIONS
Compareitem specifications
Gas Type Max Flow Rate (LPM) Wetted Materials
Availability Pricing
Aalborg Digital mass Flowmeter, Aluminum, For Air, 0 To 5 mL/min
Buy More Save More
EW-33400-00 Mfr # XFM17A-VADL9A5-00-AIR/N2
COMPARE
Air/Nitrogen/Carbon Monoxide 0.005 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, Aluminum, For Air, 0 To 20 mL/min
Buy More Save More
EW-33400-02 Mfr # XFM17A-VADL9A5-02-AIR/N2
COMPARE
Air/Nitrogen/Carbon Monoxide 0.02 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, Aluminum, For Air, 0 To 200 mL/min
Buy More Save More
EW-33400-05 Mfr # XFM17A-VADL9A5-05-AIR/N2
COMPARE
Air/Nitrogen/Carbon Monoxide 0.2 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, Aluminum, For O2, 0 To 5 mL/min
Buy More Save More
EW-33400-10 Mfr # XFM17A-VADL9A5-00-O2
COMPARE
Oxygen (O2) 0.005 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, Aluminum, For Ar, 0 To 500 mL/min
Buy More Save More
EW-33400-36 Mfr # XFM17A-VADL9A5-06-AR
COMPARE
Argon (Ar) 0.5 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, Aluminum, For Ar, 0 To 1 L/min
Buy More Save More
EW-33400-37 Mfr # XFM17A-VADL9A5-07-AR
COMPARE
Argon (Ar) 1 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, Aluminum, For He, 0 To 100 mL/min
Buy More Save More
EW-33400-44 Mfr # XFM17A-VADL9A5-04-HE
COMPARE
Helium (He) 0.1 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, Aluminum, For Co2, 0 To 20 mL/min
Buy More Save More
EW-33400-52 Mfr # XFM17A-VADL9A5-02-CO2
COMPARE
Carbon Dioxide (CO2) 0.02 Aluminum, Anodized Brass, Viton
$852.00USD / Each
Aalborg Digital mass Flowmeter, SS, For Air, 0 To 200 mL/min
Buy More Save More
EW-33401-05 Mfr # XFM17S-VADL9A5-05-AIR/N2
COMPARE
Air/Nitrogen/Carbon Monoxide 0.2 Stainless Steel, Viton
$1,209.00USD / Each
Aalborg Digital mass Flowmeter, SS, For O2, 0 To 20 mL/min
Buy More Save More
EW-33401-12 Mfr # XFM17S-VADL9A5-02-O2
COMPARE
Oxygen (O2) 0.02 Stainless Steel, Viton
$1,209.00USD / Each
Aalborg Digital mass Flowmeter, SS, For O2, 0 To 50 mL/min
Buy More Save More
EW-33401-13 Mfr # XFM17S-VADL9A5-03-O2
COMPARE
Oxygen (O2) 0.05 Stainless Steel, Viton
$1,209.00USD / Each
Aalborg Digital mass Flowmeter, SS, For O2, 0 To 200 mL/min
Buy More Save More
EW-33401-15 Mfr # XFM17S-VADL9A5-05-O2
COMPARE
Oxygen (O2) 0.2 Stainless Steel, Viton
$1,209.00USD / Each
Aalborg Digital mass Flowmeter, SS, For H2, 0 To 10 L/min
Buy More Save More
EW-33401-29 Mfr # XFM17S-VADL9A5-10-H2
COMPARE
Hydrogen (H2) 10 Stainless Steel, Viton
$1,209.00USD / Each
Aalborg Digital Mass Flowmeter, Aluminum, For Oxygen, 0 to 20 L/min
Buy More Save More
EW-33403-07 Mfr # XFM37A-VADL6-A2-20-O2
COMPARE
Oxygen (O2) 20 Aluminum, Anodized Brass, Viton
$1,009.00USD / Each
Aalborg Digital Mass Flowmeter, Aluminum, For Oxygen, 0 to 100 L/min
Buy More Save More
EW-33403-13 Mfr # XFM47A-VDDL6-A2-100-O2
COMPARE
Oxygen (O2) 100 Aluminum, Anodized Brass, Viton
$1,209.00USD / Each
Aalborg Digital Mass Flowmeter, Aluminum, For Ar, 0 to 50 L/min
Buy More Save More
EW-33403-24 Mfr # XFM37A-VADL6-A2-50-AR
COMPARE
Argon (Ar) 50 Aluminum, Anodized Brass, Viton
$1,009.00USD / Each
Aalborg Digital Mass Flowmeter, Aluminum, For He, 0 to 40 L/min
Buy More Save More
EW-33403-30 Mfr # XFM37A-VADL6-A2-40-HE
COMPARE
Helium (He) 40 Aluminum, Anodized Brass, Viton
$1,009.00USD / Each
Aalborg Digital Mass Flowmeter, Aluminum, For He, 0 to 50 L/min
Buy More Save More
EW-33403-31 Mfr # XFM37A-VADL6-A2-50-HE
COMPARE
Helium (He) 50 Aluminum, Anodized Brass, Viton
$1,009.00USD / Each
Aalborg Digital Mass Flowmeter, Stainless steel, For Ar, 0 to 60 L/min
Buy More Save More
EW-33403-67 Mfr # XFM47S-VDDL6-A2-60-AR
COMPARE
Argon (Ar) 60 Stainless Steel, Viton
$1,604.00USD / Each
Aalborg Digital Mass Flowmeter, Stainless steel, For Ar, 0 to 100 L/min
Buy More Save More
EW-33403-69 Mfr # XFM47S-VDDL6-A2-100-AR
COMPARE
Argon (Ar) 100 Stainless Steel, Viton
$1,604.00USD / Each
Showing 1 - 20 of 149