This product is now discontinued, View Related Products
Digital Hydrometer 59912
Digital specific-gravity tester with downloading capabilities
  • Measures specific gravity and temperature of liquids
  • Simply immerse the sampling tube, pull the trigger, and read the final result
  • Automatic temperature compensation with user-defined correction
  • Internal sampling pump with adjustable sampling speed
  • Display results in density, specific gravity, API degrees, Brix, % alcohol, % H2SO4, °Baume, °Plato, Proof, or a user-defined unit
  • Stores 1,100 results internally
  • Download stored results to a printer or PC using the infrared interface (sold separately)
This unit offers accurate, temperature-compensated, specific gravity results. Select from multiple units of measure to suit your measurement needs. Plain-language interface makes setup easy. Store 1,100 results internally; download stored results to a printer or PC with the optional infrared/RS-232 adapter. Optional rubber boot helps protect the meter in tougher environments.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Mettler Toledo Densito 30 PX Digital Hydrometer, infrared/RS-232 adapter EW-25751-35 Mfr # DENSITO 30PX
COMPARE
Digital Hydrometer, infrared/RS-232 adapter
Call For Price
Mettler Toledo Infrared / RS-232 adapter for Digital Hydrometer EW-25751-36 Mfr # 51325006
COMPARE
Infrared / RS-232 adapter for Digital Hydrometer
Call For Price
Mettler Toledo Protective Rubber Boot for Digital Hydrometer EW-25751-37 Mfr # 51324430
COMPARE
Protective Rubber Boot for Digital Hydrometer