Digital Protractor
Full 360° range Operate up to 500 hours on a single 9V battery
The Digital Protractor is a revolutionary tool that provides an immediate digital reading of all angles in a 360° range. The unit utilizes a liquid-filled sensor combined with precision measurement circuitry to provide unparalleled accuracy and stability. The precision machined aluminum frame provides a rigid, lightweight, ultra-precise platform for extremely accurate measurements. The frame is grooved along the bottom surface to facilitate use with round objects such as pipes or shafts and incorporates threaded holes on the bottom for rigid attachment if desired.
The front panel buttons can be used to capture desired readings or to designate an alternate reference surface from which subsequent angles are then measured. The unit can also be easily recalibrated in the field to factory accuracy using any available flat surface.
$259.25 - $498.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Resolution
Availability Pricing
COMPARE
0° ±10° = ±0.05°; 90° ±10° = ±0.1°; Other angles = ±0.2° 0.01°
$498.50USD / Each
COMPARE
0° ±10° = ±0.1°; 90° ±10° = ±0.1°; Other angles = ±0.2° 0.1°
$259.25USD / Each