DigiVac Vacuum Controllers
Multiple control and connectivity options suit any application
  • Configure dual setpoint relays as normally open or closed
  • Rugged plastic enclosure fits a 1/8 DIN panel cutout and converts to benchtop with included rubber feet
  • Field calibratable for "zero" and span
These vacuum gauges are rugged instruments that measure vacuum through 6 decades of pressure up to 760 Torr, and include the industry standard Varian 531 gauge tube sensor. The base unit can be used as a benchtop or panel mounted in a 1/8 DIN cutout and comes standard with a large bright LED display, RS232 output and 2 set point relays. The relays can be used to turn on and off devices such as valves at specific vacuum levels. The gauges allow you to select the units displayed: Torr, mBar or kPa.
The M201-2TCETH (07379-21) is fitted with 2 vacuum gauge tubes and Ethernet connectivity.
The M201VLC (07379-22) includes a valve and PID control to regulate the vacuum level in your system.
The M215V (07379-23) features analog output and RS-232 interface.
$1,162.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Digi-Vac M215V-INTL DigiVac 215V Vacuum Gauge; Analog output, 2 set points, RS-232 interface EW-07379-23 Mfr # M215V-INTL
COMPARE
±1 mTorr below 0.009 Torr, ±7% of reading from 0.010 to 0.099 Torr, ±10% of reading from 0.1 to 0.999 Torr, ±15% of reading from 1.0 to 9.9 Torr, ±50% of reading from 10 to 160 Torr, ±25% of reading above 160 Torr 0.001 Torr
$1,162.00USD / Each