Diisopropylamine 23187

Assay Min. 99%
$38.90 - $144.80USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Acros Organics AC149460010 Diisopropylamine 99% (1l) CAS 108-18-9 ((N-(1-Methylethyl)-2)propanamine) EW-88150-02 Mfr # AC149460010
COMPARE
108-18-9 1000 Diisopropylamine 99% (1l) CAS 108-18-9 ((N-(1-Methylethyl)-2)propanamine)
$94.50USD / Each
Acros Organics AC149460025 Diisopropylamine 99% (2.5l) CAS 108-18-9 EW-88150-03 Mfr # AC149460025
COMPARE
108-18-9 2500 Diisopropylamine 99% (2.5l) CAS 108-18-9
$144.80USD / Each
Acros Organics AC219432500 Diisopropylamine 99+% (250 mL) CAS 108-18-9 EW-88234-74 Mfr # AC219432500
COMPARE
108-18-9 250 Diisopropylamine 99 % (250 mL) CAS 108-18-9
$38.90USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat