This product is now discontinued, View Related Products
Dipstik™ Drum and Tank Samplers
No leachable fillers or plasticizers
  • No glass parts to break for added safety and protection
  • Easily identify layers through translucent material
  • Easy to clean
Sample free-flowing liquids from almost any source. Safety pour top permits safe and neat transfer of contents. Polypropylene samplers have heat welded bottom caps. Push/seal bottom snaps closed easily; opens by hand or with the safety opening tool sold separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Norwell Dipstik D8653242 PTFE Drum And Tank Sampler, 3.5'long EW-99200-00 Mfr # D8653242
COMPARE
PTFE Drum And Tank Sampler, 3.5'long
Call For Price
Norwell Dipstik D8653274 PTFE Drum And Tank Sampler, 7' Long EW-99200-02 Mfr # D8653274
COMPARE
PTFE Drum And Tank Sampler, 7' Long