Direct Coupled 303 Stainless Steel Rotary Vane Pumps
Ideal for high pressure applications—up to 240 psi
  • Pulseless flow at pressures up to 240 psi
  • Built-in bypass relief valve protects your pump from damage
  • Select from economical brass or corrosion-resistant 303 stainless steel bodied pumps
Power cord not included.
$977.75 - $1,217.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Motor hp Max Flow Rate (GPM) Power (amps)
Availability Pricing
COMPARE
1/2 2.3 8.0/4.0
$977.75USD / Each
COMPARE
3/4 4.3 11.0/5.5
$1,217.50USD / Each