This product is now discontinued, View Related Products
DispoLance Disposable Solids Samplers
Take any sample size you need
  • Open-ended handle allows product to continuously flow
  • Free-flowing powders and small-grain granules work best
  • Gamma sterilized and nonsterile versions available
This HDPE solids sampler is hollow all the way through. Simply pierce the bag and let the sample slide down the tube into the new container.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Length (in) Description
Availability Pricing
Dynalon DispoLance Hollow Solids Sampler, 60 mL, 300 mm L; 20/pk EW-33687-46 Mfr # 853933-5560
COMPARE
11.875 Hollow Solids Sampler, 60 mL, 300 mm L; 20/pk
Call For Price
Dynalon DispoLance Sterile Hollow Solids Sampler, 60 mL, 300 mm L; 20/pk EW-33687-47 Mfr # 853933-5561
COMPARE
11.875 Sterile Hollow Solids Sampler, 60 mL, 300 mm L; 20/pk
Call For Price
Dynalon DispoLance Hollow Solids Sampler, 100 mL, 500 mm L; 20/pk EW-33687-48 Mfr # 853933-5570
COMPARE
19.625 Hollow Solids Sampler, 100 mL, 500 mm L; 20/pk
Call For Price
Dynalon DispoLance Sterile Hollow Solids Sampler, 100 mL, 500 mm L; 20/pk EW-33687-49 Mfr # 853933-5571
COMPARE
19.625 Sterile Hollow Solids Sampler, 100 mL, 500 mm L; 20/pk