Disposable Methacrylate Cuvettes for UV Spectroscopy
% Transmission : maximum from 285 to 750 nm
Use these economical, single-use cuvettes to obtain consisten spectrophotometric data. PMMA cuvettes are preferred for UV range testing.
$208.00 - $258.00USD / Pkg of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
Dynalon Disposable UV cuvettes, methacrylate; 4.5 mL capacity, 500/pack
Buy More Save More
EW-06343-15 Mfr # 302235-0002
COMPARE
Methacrylate 4.5 10
$258.00USD / Pkg of 500
Dynalon Disposable UV cuvettes, methacrylate; 1.5 mL capacity, 500/pack
Buy More Save More
EW-06343-70 Mfr # 302225-0002
COMPARE
Methacrylate 1.5 10
$208.00USD / Pkg of 500