Dissolver Dispermat CN Series Dispersion Mills
Maximum performance for small batch applications
  • Obtain reproducible results — disperse and mix hard-to-wet fine powders and settling solids
  • Save on bench space — compact stand with control electronics built in
  • Electronic lift controls for precise impeller height positioning
  • High torque motor design with a separate ventilation fan — excellent operation at any speed setting
The Dispermat CN series is designed to provide maximum performance in a single, compact design. These models are ideal for pilot-plant-sized applications, as well as millbases in R&D and quality control/assurance. This series enables work with smaller volumes in the lower speed-range with high torque.
The digital display is infinitely variable from 0 to 11,000 rpm, and shows percent torque at the same time. An integral timer enables dispersion time to be preset with automatic shutoff — affording unattended operation and reproducible results.
Dispersion beaker/container must be ordered separately
$20,195.00 - $22,605.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
BYK-Gardner Dissolver Dispermat Dispersion Mill, 0.5 to 10 L Capacity; 220 VAC/50Hz EW-04201-40 Mfr # PO-1651
COMPARE
220
$22,605.00USD / Each
BYK-Gardner Dissolver Dispermat Dispersion Mill, 0.5 to 20 L Capacity; 400VAC/3-Phase EW-04201-42 Mfr # PO-1650
COMPARE
400
$20,195.00USD / Each