Parker Hannifin Domnick Hunter Air Purification Systems
Advanced airflow technology makes this the most energy efficient compressed air filtration system available
  • Prefilters and microfilters feature an internal float drain that eases installation and saves initial cost
  • Incident monitor (included on most models) alerts you to high differential pressure and need for filter element change
  • Housing is fully corrosion protected and carries a 10-year warranty
These modular filters are the high-efficiency answer to your compressed air filtration needs. The filter housing is Alocrom-treated aluminum with a high-quality dry powder epoxy finish. Housings carry a 10-year warranty under normal operating conditions. These filters meet or exceed the latest International ISO 8573.1 air quality standard.
Prefilters are deep bed pleated oleophobic nanofiber delivering particle removal down to 1 micron — including water and oil aerosols. Maximum remaining oil aerosol content is 0.5 ppm at 70°F (0.6 mg/m3 at 21°C).
Microfilters are deep bed pleated oleophobic nanofiber delivering particle removal down to 0.1 micron — including water and oil aerosols. Maximum remaining oil aerosol content is 0.1 ppm at 70°F (0.01 mg/m3 at 21°C).
Activated Carbon Filters use a deep-bed activated carbon element to adsorb vapors, gaseous hydrocarbons, and odors; cartridges also enhance residual oil removal when used in combination with microfilters. Maximum remaining oil vapor content is 0.003 ppm at 70°F (0.003 mg/m3 at 21°C).
Use optional mounting clamps to join adjacent filters mounted in series. Mounting clamp also functions as a wall-mounting bracket to minimize associated hardware.
$89.00 - $278.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
Compareitem specifications
Capacity
Availability Pricing
Parker Hannifin 010AO Air Prefilter Element, 21 SCFM EW-29587-01 Mfr # 010AO
COMPARE
$89.00USD / Each
Parker Hannifin 015AO Compressed Air Prefilter Element, 42 SCFM EW-29587-03 Mfr # 015AO
COMPARE
$122.00USD / Each
Parker Hannifin 020AO Compressed Air Prefilter Element, 64 SCFM EW-29587-05 Mfr # 020AO
COMPARE
$168.00USD / Each
Parker Hannifin 025AO Compressed Air Prefilter Element, 127 SCFM EW-29587-07 Mfr # 025AO
COMPARE
$202.00USD / Each
Parker Hannifin 030AO Compressed Air Prefilter Element, 233 SCFM EW-29587-09 Mfr # 030AO
COMPARE
$278.00USD / Each
Parker Hannifin 010AA Compressed Air Microfilter Element, 21 SCFM EW-29587-11 Mfr # 010AA
COMPARE
$95.00USD / Each
Parker Hannifin 015AA Compressed Air Microfilter Element, 42 SCFM EW-29587-13 Mfr # 015AA
COMPARE
$122.00USD / Each
Parker Hannifin 020AA Compressed Air Microfilter Element, 64 SCFM EW-29587-15 Mfr # 020AA
COMPARE
$184.00USD / Each
Parker Hannifin 025AA Compressed Air Microfilter Element, 127 SCFM EW-29587-17 Mfr # 025AA
COMPARE
$202.00USD / Each
Parker Hannifin 030AA Compressed Air Microfilter Element, 233 SCFM EW-29587-19 Mfr # 030AA
COMPARE
$278.00USD / Each
Parker Hannifin 010ACS Compressed Air Activated Carbon Filter Element, 21 SCFM EW-29587-21 Mfr # 010ACS
COMPARE
$89.00USD / Each
Parker Hannifin 015ACS Compressed Air Activated Carbon Filter Element, 42 SCFM EW-29587-23 Mfr # 015ACS
COMPARE
$113.00USD / Each
Parker Hannifin 020ACS Compressed Air Activated Carbon Filter Element, 64 SCFM EW-29587-25 Mfr # 020ACS
COMPARE
$168.00USD / Each
Parker Hannifin 025ACS Compressed Air Activated Carbon Filter Element, 127 SCFM EW-29587-27 Mfr # 025ACS
COMPARE
$202.00USD / Each
Parker Hannifin 030ACS Compressed Air Activated Carbon Filter Element, 233 SCFM EW-29587-29 Mfr # 030ACS
COMPARE
$272.00USD / Each
Showing 1- 15 of 15