Doran 7000 XL Washdown Industrial Scales
IP69K-rated display unit tilts to any angle for easy viewing; mount up to 10 feet away
  • Water and corrosion resistant—electronics and sensor are enclosed inside the heavy-duty stainless steel case
  • Quad Spring™ system provides a 1000% overload capacity to virtually eliminate damage from shock
  • Calibrate at 100%, 50%, 25%, or 10% of capacity
  • Update rate from 1 to 10 readings per second
$1,210.00 - $1,345.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
Doran Scale 7005XL Washdown Industrial Scale, 5 Lb/2.3kg, 115 V EW-11910-00 Mfr # 7005XL
COMPARE
2.3 5 115
$1,210.00USD / Each
Doran Scale 7005XL-230 Washdown Industrial Scale, 5 Lb/2.3kg, 230 V EW-11910-05 Mfr # 7005XL-230VAC
COMPARE
2.3 5 230
$1,210.00USD / Each
Doran Scale 7010XL Washdown Industrial Scale, 10lb/4.5kg, 115 V EW-11910-10 Mfr # 7010XL
COMPARE
4.5 10 115
$1,210.00USD / Each
Doran Scale 7010XL-230 Washdown Industrial Scale, 10lb/4.5kg, 230 V EW-11910-15 Mfr # 7010XL/EXOPT123
COMPARE
4.5 10 230
$1,210.00USD / Each
Doran Scale 7025XL Washdown Industrial Scale, 25 Lb/11.3kg, 115 V EW-11910-20 Mfr # 7025XL
COMPARE
11.3 25 115
$1,210.00USD / Each
Doran Scale 7025XL-230 Washdown Industrial Scale, 25 Lb/11.3kg, 230 V EW-11910-25 Mfr # 7025XL-230VAC
COMPARE
11.3 25 230
$1,210.00USD / Each
Doran Scale 7050XL/12 Washdown Industrial Scale, 50lb/22.7kg, 115 V EW-11910-30 Mfr # 7050XL/12
COMPARE
22.7 50 115
$1,345.00USD / Each
Doran Scale 7050XL/12-230VAC Washdown Industrial Scale, 50lb/22.7kg, 230 V EW-11910-35 Mfr # 7050XL/12-230VAC
COMPARE
22.7 50 230
$1,345.00USD / Each
Doran Scale 7100XL/12 Washdown Industrial Scale, 100lb/45kg, 115 V EW-11910-40 Mfr # 7100XL/12
COMPARE
45 100 115
$1,345.00USD / Each
Doran Scale 7100XL/12-230VAC Washdown Industrial Scale, 100lb/45kg, 230 V EW-11910-45 Mfr # 7100XL/12-230VAC
COMPARE
45 100 230
$1,345.00USD / Each
Showing 1- 10 of 10