Double-Walled Heat-Shrink Tubing, PTFE
Ideal for chemically inert applications
  • Global market leader in range of formulations for most extensive chemical compatibility
Double-walled PTFE tubing combines an exterior of heat-shrink PTFE with an inner layer of FEP to provide a tight, moisture-proof bond. Comes in 4-ft lengths; 3 lengths per pack, for a total of 12 ft/pk. Opaque.
$52.00 - $396.00USD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubing ID (in)
Availability Pricing
COMPARE
1/16
$52.00USD / Pkg of 1
COMPARE
7/64
$115.00USD / Pkg of 1
COMPARE
3/16
$396.00USD / Pkg of 1
COMPARE
3/16
$107.00USD / Pkg of 1
COMPARE
15/64
$116.00USD / Pkg of 1
COMPARE
11/32
$267.00USD / Pkg of 1