Dow Corning® Pharma-65 Tubing
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing.
Characteristics: Platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex, or other organic additives. Remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicon to silicone tubing. Durometer shore A rating of 62.
Certification: Elastomer is FDA compliant and meets USP Class VI requirements. Meets 3A Sanitary Standards and European Pharmacopoeia (EP) requirements. Tubing is manufactured according to Bulk Pharmaceutical (BP) and current GMP. Temperature range: –58 to 482°F (–50 to 250°C).
$168.00 - $754.00USD / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Dow Corning Pharma-65 Silicone Tubing, 1/8" x  1/4"; 50 ft/Pk EW-96107-00 Mfr # 4090602
COMPARE
19 1/8 1/4
$168.00USD / Pkg of 1
Dow Corning Pharma-65 Silicone Tubing, 3/16" x 5/16"; 50 ft/Pk EW-96107-01 Mfr # 4041047
COMPARE
13 3/16 5/16
$206.00USD / Pkg of 1
Dow Corning Pharma-65 Silicone Tubing, 3/16" x 3/8"; 50 ft/Pk EW-96107-02 Mfr # 4077179
COMPARE
15 3/16 3/8
$318.00USD / Pkg of 1
Dow Corning Pharma-65 Silicone Tubing, 1/4" x 7/16"; 50 ft/Pk EW-96107-04 Mfr # 4094222
COMPARE
20 1/4 7/16
$324.00USD / Pkg of 1
Dow Corning Pharma-65 Silicone Tubing, 1/4" x 1/2"; 50 ft/Pk EW-96107-05 Mfr # 4008334
COMPARE
19 1/4 1/2
$520.00USD / Pkg of 1
Dow Corning Pharma-65 Silicone Tubing, 3/8" x 5/8"; 50 ft/Pk EW-96107-09 Mfr # 4008335
COMPARE
13 3/8 5/8
$619.00USD / Pkg of 1
Dow Corning Pharma-65 Silicone Tubing, 1/2" x 3/4"; 50 ft/Pk EW-96107-11 Mfr # 4008336
COMPARE
11 1/2 3/4
$754.00USD / Pkg of 1
Showing 1- 7 of 7