Dow Corning Pharma-80 Tubing
Applications: Pharmaceutical, biotechnology, and ultra-pure fluid processing.
Characteristics: Semi-transparent platinum-cured silicone tubing with a very low extractable profile; it contains no phthalates, plasticizers, latex or other organic additives. Pharma tubing is hydrophobic, and remains flexible and resilient for long service life. Dow Corning maintains complete traceability from silicone to silicone tubing. Durometer, shore A rating of 75.
Certification: Elastomer exceeds test guidelines for USP Class VI Plastics; tubing itself is FDA compliant, and meets 3A Sanitary Standards and European Pharmacopoeia (EP) Requirements. Pharma tubing is produced under Bulk Pharmaceutical (BP) current GMP principals.
<
$276.00 - $1,288.00USD / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Pressure (PSI) Tubing ID (in) Tubing OD (in)
Availability Pricing
Dow Corning Pharma-80 Tubing, 1/8" (3.2) x 1/4" (6.4), 50-ft pk EW-96109-00 Mfr # 4090082
COMPARE
249 1/8 1/4
$276.00USD / Pkg of 1
Dow Corning Pharma-80 Tubing, 3/16" (4.8) x 5/16" (8.0), 50-ft pk
Always in Stock
EW-96109-01 Mfr # 4090084
COMPARE
249 3/16 5/16
$315.00USD / Pkg of 1
Dow Corning Pharma-80 Tubing, 1/4" (6.4) x 3/8" (9.6), 50-ft pk EW-96109-03 Mfr # 4066764
COMPARE
249 1/4 3/8
$351.00USD / Pkg of 1
Dow Corning Pharma-80 Tubing, 1/4" (6.4) x 1/2" (12.8), 50-ft pk EW-96109-05 Mfr # 4008432
COMPARE
260 1/4 1/2
$878.00USD / Pkg of 1
Dow Corning Pharma-80 Tubing, 3/8" (9.6) x 5/8" (16.0), 50-ft pk EW-96109-09 Mfr # 3100651
COMPARE
231 3/8 5/8
$935.00USD / Pkg of 1
Dow Corning Pharma-80 Tubing, 1/2" (12.8) x 3/4" (19.2), 50-ft pk
Buy More Save More
EW-96109-11 Mfr # 4008433
COMPARE
200 1/2 3/4
$1,288.00USD / Pkg of 1