This product is now discontinued, View Related Products

Dowex(r) 50WX8-100 ion-exchange resin

Call For Price

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
CAS Number Weight (g) Description
Availability Pricing
Acros Organics AC335331000 50WX8-100 ion-exchange resin (100g) EW-88305-08 Mfr # AC335331000
COMPARE
11119-67-8 100 50WX8-100 ion-exchange resin (100g)
Call For Price
Dowex AC335335000 50WX8-100 ion-exchange resin (500g) EW-88305-09 Mfr # AC335335000
COMPARE
11119-67-8 500 50WX8-100 ion-exchange resin (500g)
Call For Price
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat