This product is now discontinued, View Related Products

Drager Pac 3500/5500 Single-Gas Monitors

Three alarms functions – audible, visual, and vibration – make alarm conditions clear
The monitor features a robust, rubber-coated weatherproof polymer housing rated to IP65 that will stand up to the typical abuse in an industrial environment. Easily operated using just two buttons. Its small size makes it comfortable and easy to wear all day on a belt, pocket, or lapel. A loud audible alarm, two bright visual alarms, and strong vibrating alarm alert you to hazardous concentrations of gas. Built-in event logger stores up to 60 alarm activations or other events.
6 PRODUCT OPTIONS
Compareitem specifications
Battery Type Gases Measured
Availability Pricing
Drager PAC 3500 Single-Gas Monitor, O2, 0 to 25% EW-86515-01 Mfr # 4543959
COMPARE
Nonreplaceable lithium batteries O2
Call For Price
Drager PAC 3500 Single-Gas Monitors, CO, 0 to 500 ppm EW-86515-03 Mfr # 4543957 8322001
COMPARE
Nonreplaceable lithium batteries CO
Call For Price
Drager PAC 3500 Single-Gas Monitors, H2S EW-86515-05 Mfr # 4543958
COMPARE
Nonreplaceable lithium batteries H2S
Call For Price
Drager PAC 5500 Single-Gas Monitors, O2, 0 to 25% EW-86515-07 Mfr # 4543962
COMPARE
Rechargable battery O2
Call For Price
Drager PAC 5500 Single-Gas Monitors, CO, 0 to 500 ppm EW-86515-09 Mfr # 4543960
COMPARE
Rechargable battery CO
Call For Price
Drager PAC 5500 Single-Gas Monitors, H2S EW-86515-11 Mfr # 4543961
COMPARE
Rechargable battery H2S
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat