This product is now discontinued, View Related Products
Druck Portable Digital Pressure Indicator (DPI) with Alarm
  • Audiovisual alarm alerts you once high setpoint is reached--perfect for maintenance and trouble shooting.
  • NEMA 12(IP54) case--ideal for field use.
  • View minimum and maximum readings as well as ambient temperature.
  • Recalibrate using zero and TARE keys.
  • Automatic shut-off and low-battery indication.
  • Perform a one-minute LEAK test with count-down timer displayed.
Call For Price
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
GE Druck DPI 705 Portable Pressure Indicator with Internal Sensor, 0 to 100 psig; 1/8" NPT(F) EW-68452-99 Mfr # DPI 705 100PSIG
COMPARE
±0.1% full-scale 100 0.01 psi
Call For Price
GE Druck DPI 705R Portable Pressure Indicator with Internal Sensor, 0 to 500 psig; 1/8" NPT(F) EW-68453-00 Mfr # I705R-500PSIG
COMPARE
±0.1% full-scale 500
Call For Price
GE Druck DPI 705R Portable Pressure Indicator with Remote Sensor, 0 to 100 psia; 1/8" NPT(F) EW-68453-44 Mfr # I705R-100PSIG
COMPARE
±0.1% full-scale 0.01 psi
Call For Price
GE Druck DPI 705 Portable Pressure Indicator with Internal Sensor, 0 to 3 psid; 1/8" NPT(F) EW-68970-40 Mfr # I705-3PSID
COMPARE
±0.1% full-scale 0.0001 psi
Call For Price
GE Druck DPI 705 Portable Pressure Indicator with Internal Sensor, 0 to 30 psig; 1/8" NPT(F) EW-68970-50 Mfr # I705-30PSIG
COMPARE
±0.1% full-scale 30 0.001 psi
Call For Price
GE Druck DPI 705 Portable Pressure Indicator with Internal Sensor, 0 to 30 psia; 1/8" NPT(F) EW-68970-60 Mfr # I705-30PSIA
COMPARE
±0.1% full-scale 0.001 psi
Call For Price
GE Druck DPI 705 Portable Pressure Indicator with Internal Sensor, 0 to 300 psig; 1/8" NPT(F) EW-68970-70 Mfr # I705-300PSIG
COMPARE
±0.1% full-scale 300 0.01 psi
Call For Price
Showing 1- 7 of 7