Drummond Pipet-Aid® Original Portable Pipette Controller
Take wherever needed in the lab—cordless and rechargeable
  • Pipet over 2000 times on a single charge
  • Flow rate better than 25 mL at 5 seconds with 100 mL pipet
Drummond Pipet-Aid® Original Portable Pipette Controller features a built-in pump and provides cordless usability, making it ideal for use in the lab or field. Pipette controllers are designed to transfer liquids with the use of serological, volumetric, and blow-out pipets in either glass, plastic, or something similar from 1 to 100 mL.
Operates on a rechargeable nickel metal hydride (NiMH) battery. Use continuously for two hours before recharging is required. Can be used while recharging.
$470.50 - $529.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Dispensing (mL) Min Dispensing (mL) Power (VAC)
Availability Pricing
Drummond Pipet-Aid® Original Portable Pipette Controller; 120 VAC, 60 Hz EW-07897-30 Mfr # 4-000-100
COMPARE
100 1 120
$470.50USD / Each
Drummond Pipet-Aid® Original Portable Pipette Controller; 220 VAC, 50 Hz EW-07897-32 Mfr # 4-000-200
COMPARE
100 1 220
$529.50USD / Each