Dry-Keeper Desiccator Cabinets — Electric
Nonvacuum desiccators available in non-electric or electronic models
  • Horizontal or vertical orientations
  • Gasketed doors for an airtight seal
  • Transparent sides and top allow easy identification of inner contents without opening up the door
Electronic Auto-Desiccator Cabinets feature a permanent desiccant which automatically regenerates for worry-free storage or drying. The horizontal and vertical models are made of clear acrylic and have the desiccant located in the control unit; the desiccant is automatically regenerated by a 30-minute cycle every five hours to lower humidity to 30 to 40%. The Dry-Keeper Plus vertical cabinet is made from nontransparent, insulated ABS plastic and has an enclosed permanent desiccant; the electronic controller lowers relative humidity inside the cabinet to 20 to 30% in six hours (based on unloaded, continuous operation).
$2,324.00 - $2,387.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Exterior Height (in) Exterior Width (in) Orientation
Availability Pricing
Dry-Keeper H420580003 Desiccator Cabinets, Horizontal; Electronic EW-08909-60 Mfr # H420580003
COMPARE
13.75 24.5 Horizontal
$2,357.00USD / Each
Dry-Keeper H420561003 Desiccator Cabinets, Vertical; Electronic EW-08909-62 Mfr # H420561003
COMPARE
20 12.5 Vertical
$2,387.00USD / Each
Dry-Keeper H420610000 Plus Desiccator Cabinets, Vertical; Electronic EW-08909-64 Mfr # H420610000
COMPARE
13.875 12 Vertical
$2,324.00USD / Each