Dual-Feed Systems with Individual Timer Controls
Simple, compact, and easy-to-use panel allows user to positions timer and pumps for optimal flexibiity
  • Flexible dual-pump configuration
  • 0.6 to 6.3 GPH (2.2 to 23.8 LPH) outputs up to 150 psi (10.3 bar)
  • Individual 24/7 timers for each pump
  • Simple "plug and play" installation — just connect fluid inlet and outlet and power
This simple, economical system provides dual individual timers and pump receptacles in a wall-mount configuration. This is an ideal economical option for dual biocide feed applications. The systems come with two Walchem pumps; call for information on substituting another brand of pump.
$3,120.00 - $3,820.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Max Pressure (PSI) Power (VAC)
Availability Pricing
COMPARE
0.6 10.3 115
$3,120.00USD / Each
COMPARE
2 7.2 115
$3,590.00USD / Each
COMPARE
4.3 3.4 115
$3,690.00USD / Each
COMPARE
6.3 2.06 115
$3,820.00USD / Each
COMPARE
0.6 10.3 230
$3,120.00USD / Each
COMPARE
2 7.2 230
$3,590.00USD / Each
COMPARE
4.3 3.4 230
$3,690.00USD / Each
COMPARE
6.3 2.06 230
$3,820.00USD / Each
Showing 1- 8 of 8
Need Help with this product?
Call Us At1-847-549-7600
Live chat