Dual-Layer Gradient-Density Catridges
Dual-layer design has a holding capacity of two to three times traditional cartridges
Dual-layer cartridges are high-capacity cartridges that combine a prefiltration layer with a selective postfiltration layer. Each layer is a separate gradient-density filter, which reduces larger particles on the outer layers and finer particles on the inner layers. Ideal for removing a broad range of deformable and nondeformable particles such as sand, silt, sludge, rust and scale.
This design has a holding capacity that is two to three times traditional spun or wound designs. Catridges are made of bacteria and chemical-resistant polypropylene; suitable to 145°> F (63°>C). These cartridges are certified to NSF Standard 42 for materials.
$23.75 - $57.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Length (in) Micron Rating
Availability Pricing
Pentair 155360-43 20" dual-gradient density polypropylene cartridge, 25 µm prefilter, 1 µm EW-29802-20 Mfr # 155360-43
COMPARE
20 1
$57.00USD / Each
Pentair 155358-43 20" dual-gradient density polypropylene cartridge, 50 µm prefilter, 5 µm EW-29802-21 Mfr # 155358-43
COMPARE
20 5
$49.50USD / Each
Pentair 155356-43 20" dual-gradient density polypropylene cartridge, 75 µm prefilter, 25 µm
Always in Stock
EW-29802-22 Mfr # 155356-43
COMPARE
20 75
$53.50USD / Each
Pentair 155359-43 10" dual-gradient density polypropylene cartridge, 25 µm prefilter, 1 µm EW-29802-23 Mfr # 155359-43
COMPARE
10 25
$26.50USD / Each
Pentair C155357-43 10" dual-gradient density polypropylene cartridge, 50 µm prefilter, 5 µm EW-29802-24 Mfr # C155357-43
COMPARE
10 5
$23.75USD / Each
Pentair 155355-43 10" dual-gradient density polypropylene cartridge, 75 µm prefilter, 25 µm EW-29802-25 Mfr # 155355-43
COMPARE
10 25
$24.75USD / Each
Showing 1- 6 of 6
Need Help with this product?
Call Us At1-847-549-7600
Live chat