Dual-Point Level Sensor Kits
Kits come with all you need to mount two sensors—ideal for drain/fill operations
Create sensors from 10" to 36" long
This sensor kit can be configured and reconfigured to fit different applications. Install your dual-level sensor in any 1/2" or 2" NPT(F) bung opening. Switches are rated for 60 W; position floats for either NO or NC operation.
Slide floats onto the outside of the switch segments. Attach additional 1/2" OD segments or cut to the length you need. Connectors can act as stops to hold floats. Each kit includes a 9"L mounting segment, two 4-1/2"L switch segments, two 12"L blank segments, two floats, and all necessary connectors.
$410.00 - $751.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Depth (ft) Max Pressure (PSI) Min Depth (ft)
Availability Pricing
Gems Dual-Point Brass/Buna N Level Sensor Kit EW-07188-95 Mfr # ML-4444
COMPARE
100
$410.00USD / Each
Gems ML8888 Polypropylene Dual Float Level Switch Kit EW-07188-96 Mfr # ML-8888
COMPARE
150
$631.00USD / Each
Gems ML5555 Stainless Steel Dual Float Level Switch Kit EW-07188-97 Mfr # ML-5555
COMPARE
200
$751.00USD / Each