Dwyer Air Velocity Transmitter
This air velocity transmitter is the ideal instrument for monitoring air flow. This transmitter uses a heated mass flow sensor, which allows for precise velocity measurements at various flow rates and temperatures. The transmitter’s 16 field-selectable ranges allow the versatility to be selected for several airflow applications. The LED display version produces a complete, low-cost solution for local indication of airflow.
FEATURES
  • Ranges to 15,000 FPM or 75 MPS
  • Optional bright LED display
  • Easy push button set-up
  • Compact housing
  • 4-20mA output
  • Digital filter for signal damping
ITEM(S) ON SALE
$484.50 - $607.75USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Dwyer 641-6-LED Transmitter, 6" probe length, LED display EW-10778-74 Mfr # 641-6-LED
COMPARE
Transmitter, 6" probe length, LED display
$602.65USD / Each
$709.00USD / Each
Dwyer 641RM-12-LED Transmitter, 6" remote mounted probe, 6' cable and LED display EW-10778-75 Mfr # 641RM-12-LED
COMPARE
Transmitter, 6" remote mounted probe, 6' cable and LED display
$607.75USD / Each
$715.00USD / Each
Dwyer 641-12-LED Transmitter, 12" probe length, LED display EW-10778-77 Mfr # 641-12-LED
COMPARE
Transmitter, 12" probe length, LED display
$602.65USD / Each
$709.00USD / Each
Dwyer 641-18 Transmitter 18" probe length, no display EW-10778-78 Mfr # 641-18
COMPARE
Transmitter 18" probe length, no display
$484.50USD / Each
$570.00USD / Each