Dwyer Digihelic Differential Pressure Controllers
  • FEATURES
  • Field programmable for pressure, velocity or flow operation
  • 4-20mA process output
  • 0.5% full scale accuracy sensor
  • Modbus communications
  • 1/8 DIN housign with NEMA 4X (IP66) front face
The Digihelic® Differential Pressure Controller is the ideal instrument for pressure, velocity and flow applications, achieving a 0.5% full scale accuracy on ranges from 5 to 100 in. w.c. It provides a 4-20mA process output, 2 SPDT relays with adjustable dead bands, and selection of pressure, velocity or flow operation. Units operate from either 120/220 VAC or 24 VDC.
$498.50 - $648.75USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Dwyer DH-002 0.25 In W.C. Differential Pressure Controller EW-68437-17 Mfr # DH-002
COMPARE
±0.5% full-scale
$551.50USD / Each
Dwyer DH-004 1.000 In W.C. Differential Pressure Controller EW-68437-18 Mfr # DH-004
COMPARE
±0.5% full-scale
$648.75USD / Each
Dwyer DH-006 Digihelic Differential Pressure Controller, 5" WC EW-68437-19 Mfr # DH-006
COMPARE
±0.5% full-scale
$498.50USD / Each
Dwyer DH-007 Digihelic Differential Pressure Controller, 10" WC EW-68437-20 Mfr # DH-007
COMPARE
±0.5% full-scale
$498.50USD / Each
Dwyer DH-008 Digihelic Differential Pressure Controller, 25" WC EW-68437-21 Mfr # DH-008
COMPARE
±0.5% full-scale
$498.50USD / Each
Dwyer DH-009 Digihelic Differential Pressure Controller, 50" WC EW-68437-22 Mfr # DH-009
COMPARE
±0.5% full-scale
$498.50USD / Each
Dwyer DH-010 100 In Wc Diff Press Controller Digihelic EW-68437-23 Mfr # DH-010
COMPARE
±0.5% full-scale
$498.50USD / Each
Showing 1- 7 of 7