Dwyer Mark II Inclined Liquid Manometer
Pressure measurement for stationary and portable applications
  • ±3% accuracy
  • Gauge fluid included
Inclined liquid manometers are capable of pressure measurements above and below atmospheric as well as differential pressure measurements, and are economically priced, compact and available in both stationary and portable configurations. Construction is simple with virtually indestructible molded white styrene-acrylonitrile housing, indicating tube and fluid wells, molded ABS knobs and zero adjust plunger, shock mounted glass level vial and leak proof 'O' ring seals. These manometers are very easy to use - simply fill the reservoir, adjust fluid level to zero, connect the tubing, and the gauge is ready for operation.
Curved inclined-vertical tube of model 68062-58 provides higher ranges with more easily read increments at low readings and is excellent for general purpose use.
$37.50 - $97.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Dwyer MARK II 25 Inclined Manometer, 3" WC EW-68062-58 Mfr # MARK II 25
COMPARE
±3% full-scale
$37.50USD / Each
Dwyer MARK II 40-1 Inclined Manometer, 1" WC EW-68062-60 Mfr # MARK II 40-1
COMPARE
±3% full-scale
$97.00USD / Each