Dwyer Mini-Photohelic Differential Gauges - Switches
  • FEATURES
  • Ranges to 100 inches of water
  • Two 5A SPDT relay set points
  • LED indicators for switch status
  • Easy push button set-up
  • Compact housing
The mini-photohelic differential pressure gauge/switch combines a differential pressure gauge with two SPDT switch set points. It is designed to measure and control positive, negative or differential pressures consisting of non-combustible and non-corrosive gases. The gauge reading is independent of the switch operation. The switching status is visible by LED indicators located on the front and rear of the gauge. The set points are adjusted with pushbuttons on the back of the unit. This extremely compact switch/gauge is ideal for fume hoods, dust collection, pneumatic conveying and clean room applications.
ITEM(S) ON SALE
$249.90USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Dwyer MP-000 0-0.5 Inh20 Mini-Photoheli Press Gauge/Switch EW-68462-82 Mfr # MP-000
COMPARE
±5% full-scale at 70°F (21°C) 0.01" WC
$249.90USD / Each
$294.00USD / Each
Dwyer MP-001 0-1.0 Inh20 Mini-Photoheli Diff Press Gauge/Switch EW-68462-83 Mfr # MP-001
COMPARE
±5% full-scale at 70°F (21°C) 0.02" WC
$249.90USD / Each
$294.00USD / Each
Dwyer MP-002 0-2.0 Inh20 Mini Photoheli Press Gauge/Switch EW-68462-84 Mfr # MP-002
COMPARE
±5% full-scale at 70°F (21°C) 0.05" WC
$249.90USD / Each
$294.00USD / Each
Dwyer MP-003 0-3.0 Inh20 Mini Photoheli Diff Press Gauge/Switch EW-68462-85 Mfr # MP-003
COMPARE
±5% full-scale at 70°F (21°C) 0.1" WC
$249.90USD / Each
$294.00USD / Each
Dwyer MP-005 0-5.0 Inh20 Mini Photoheli Diff Press Gauge/Switch EW-68462-86 Mfr # MP-005
COMPARE
±5% full-scale at 70°F (21°C) 0.1" WC
$249.90USD / Each
$294.00USD / Each
Dwyer MP-010 Mini Photohelic Diff Press Gau 0-10 Inh20 EW-68462-87 Mfr # MP-010
COMPARE
±5% full-scale at 70°F (21°C) 0.2" WC
$249.90USD / Each
$294.00USD / Each