Dyna-Lume Halogen Illuminators

High intensity light for your inspection applications
  • Fully posable goosenecks allow for multiple light configurations
These quartz-halogen illuminators are available with general-purpose, high-performance, or high-intensity illuminators in single- and dual-head configurations. Order the optional magnifier for convenient, close-up inspections. Optional infrared heat shields protect heat-sensitive samples — ideal for biological applications.
$275.30 - $368.20USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bulb Type Power (VAC) Description
Availability Pricing
Scientific Instruments 240-160 Dual-head General-Purpose Illuminators; 115V EW-09809-55 Mfr # 240-160
COMPARE
Combination/64º 115 Dual-head General-Purpose Illuminators; 115V
$275.30USD / Each
Scientific Instruments 240-250 Dual-head High-performance Illuminators; 115V EW-09809-62 Mfr # 240-250
COMPARE
Spot/15º or Flood/50º 115 Dual-head High-performance Illuminators; 115V
$312.30USD / Each
Scientific Instruments 240-352 Dual-head, High-intensity Illuminators, 115V EW-41600-30 Mfr # 240-352
COMPARE
Combination/15º, Flood/30º, or Spot/9º 115 Dual-head, High-intensity Illuminators, 115V
$368.20USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat