Eagle Tower Flammable Storage Cabinets
Include 4" (10.2 cm) high adjustable legs to elevate your cabinet
Individually adjustable legs allow you to raise the cabinet for placing over floor ventilation, protect the cabinet where moisture is a concern, easily clean underneath, and move the cabinet with a forklift or pallet jack. Constructed of epoxy-coated, double-walled, 18-gauge steel with 1-1/2" (3.8 cm) insulating barrier — cabinets are ideal for safely storing flammable liquid containers up to 5 gallons (19 L) in size. Cabinets feature dual vents, grounding attachments, and key lock. Include adjustable HDPE shelf trays for excellent spill containment. Hazard label: FLAMMABLE — KEEP FIRE AWAY.
$970.25 - $2,756.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal) Self-Closing
Availability Pricing
Eagle 1925LEGS Tower Flammable Safety Cabinet, 12 gallon, manual-latching door EW-09418-41 Mfr # 1925LEGS
COMPARE
12 No
$1,031.00USD / Each
Eagle 1906LEGS Tower Flammable Safety Cabinet, 16 gallon, manual-latching door EW-09418-42 Mfr # 1906LEGS
COMPARE
16 No
$1,214.00USD / Each
Eagle 1932LEGS Tower Flammable Safety Cabinet, 30 gallon, manual-latching door EW-09418-43 Mfr # 1932LEGS
COMPARE
30 No
$1,410.50USD / Each
Eagle 1947LEGS Tower Flammable Safety Cabinet, 45 gallon, manual-latching door EW-09418-44 Mfr # 1947LEGS
COMPARE
45 No
$1,598.50USD / Each
Eagle 1962LEGS Tower Flammable Safety Cabinet, 60 gallon, manual-latching door EW-09418-45 Mfr # 1962LEGS
COMPARE
60 No
$1,831.50USD / Each
Eagle 1992LEGS Tower Flammable Safety Cabinet, 90 gallon, manual-latching door EW-09418-46 Mfr # 1992LEGS
COMPARE
90 No
$2,528.50USD / Each
Eagle 1924LEGS Tower Flammable Safety Cabinet, 12 gallon, self-closing door EW-09418-51 Mfr # 1924LEGS
COMPARE
12 Yes
$970.25USD / Each
Eagle 1905LEGS Tower Flammable Safety Cabinet, 16 gallon, self-closing door EW-09418-52 Mfr # 1905LEGS
COMPARE
16 Yes
$1,327.50USD / Each
Eagle 3010LEGS Tower Flammable Safety Cabinet, 30 gallon, self-closing door EW-09418-53 Mfr # 3010LEGS
COMPARE
30 Yes
$1,597.00USD / Each
Eagle 4510LEGS Tower Flammable Safety Cabinet, 45 gallon, self-closing door EW-09418-54 Mfr # 4510LEGS
COMPARE
45 Yes
$1,878.00USD / Each
Eagle 6010LEGS Tower Flammable Safety Cabinet, 60 gallon, self-closing door EW-09418-55 Mfr # 6010LEGS
COMPARE
60 Yes
$2,342.50USD / Each
Eagle 9010LEGS Tower Flammable Safety Cabinet, 90 gallon, self-closing door EW-09418-56 Mfr # 9010LEGS
COMPARE
90 Yes
$2,756.00USD / Each
Showing 1- 12 of 12