EchoTherm Digital Stirring Hot Plates
  • Temperature ramping provides exact heating and cooling rates
  • PID digital control for precise plate and probe temperatures
  • Timer allows hands-free operation
Set temperature ramp rate from 1°C/hr to 400°C/hr. Platinum RTD measurement provides precision and accuracy over entire temperature range to 842°F (450°C). Memory stored temperature calibration can be reset using the probe calibration kit (sold separately). Set timer with auto-off for exact timed heating and stirring of your sample. Stirring hot plates have an epoxy-coated cast aluminum chassis and a splash-resistant keyboard cover. Indicator lights let you know if top plate is hot or unit has reached overtemperature status. Ceramic glass top plates are impervious to chemical spills; feature a cast-aluminum frame to keep edges cool. What’s included: a 6-ft three-wire grounded AC power cord and fuse holder. Order probes separately.
$1,775.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Torrey Pines EchoTherm HS30-CP Digital Stirring Hot Plate, 115 VAC EW-51808-20 Mfr # HS30-CP
COMPARE
115
$1,775.00USD / Each