Master Appliance Master® Compact Heat Guns
Compact, light weight and easy-to-use
The heat guns are light weight and include a 3-wire grounded 6-ft cord. These heat guns also feature a protective switch guard, hanging loop, and a built-in stand for hands-free use.
Proheat heat guns are designed for use in a wide range of applications, such as heating sensitive materials including specialty connectors and shrink tubing. Featuring the ability to adjust the air temperature with a built-in temperature controller. The electronic closed loop temperature control system maintains a constant temperature and protects from overheating.
The Ecoheat heat gun is an economical solution for a variety of applications, featuring two heat and two air flow settings.
$243.25 - $367.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Air Volume (CFM) Max Temperature (° F) Power (VAC)
Availability Pricing
Master Appliance PH-1200 Proheat VariTemperature Heat Gun, 130° to 1000°F, 120V, 60Hz EW-03033-00 Mfr # PH-1200
COMPARE
16 1000 120
$243.25USD / Each
Master Appliance PH-1400 Proheat LCD Dial-in Heat Gun, 130° to 1000°F, 4-16 CFM, 120V, 60Hz EW-03033-05 Mfr # PH-1400A
COMPARE
4 - 16 1000 120
$367.25USD / Each