Economical Drives for Micropump A-Mount Pump Heads 4742
A variety of low-cost drives for numerous applications
  • Magnetic coupling minimizes friction—all horsepower is transmitted directly to the gears
  • Choose either an explosion-proof or air-powered model for safe pumping in hazardous areas
  • Hundreds of hours of dependable leakproof service
$457.00 - $1,609.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Motor RPM (rpm) Motor Enclosure Type Description
Availability Pricing
Micropump 81101 Economical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC
Always in Stock
EW-07003-90 Mfr # 81101
COMPARE
4000 PM brush, TENV Economical drive for A-Mount pump heads, 150 to 4000 rpm, 0 to 24 VDC
$457.00USD / Each
Micropump 81094 Drive for A-Mount pump heads, explosion-proof (XPRF), 3150 rpm, 115/230 VAC EW-07144-95 Mfr # 85734
COMPARE
3150 PSC Drive for A-Mount pump heads, explosion-proof (XPRF), 3150 rpm, 115/230 VAC
$1,609.00USD / Each
Micropump L22096 A-Mount Air-Driven Pump Drive, 500 to 8000 rpm, 5 to 11 scfm, 50 to 80 psi EW-07144-97 Mfr # L22096
COMPARE
8000 TENV A-Mount Air-Driven Pump Drive, 500 to 8000 rpm, 5 to 11 scfm, 50 to 80 psi
$674.25USD / Each