McMillan Flow Economical Modular Flow Rate Sensors
Ryton® PPS materials for use in aggressive and non-aggressive gas or liquid systems
  • A range of available configurations to suit most applications
  • Voltage output to monitor and record flow rates and totals
  • Interface with data acquisition system
These sensors are ideal for low-flow applications involving mildly acidic or slightly corrosive gases and liquids. Economically designed, sensors provide a single 0 to 5 VDC output signal. This single signal is ideal for simple, low-cost flow rate measurement or for integration of the sensor into an existing central control system.
Ryton PPS sensors are ideal for liquids or air. Sensors for liquids can be used with a wide variety of transparent, low-viscosity liquids under 10 cSt. Sensors measure a wide flow range from as low as 20 mL/min to as high as 500 L/min.
The wetted materials are epoxy, glass-filled polyphenylene sulfide (Ryton PPS), glass, stainless steel, sapphire, and Viton.
Note: A power supply is required for these flow sensors—order separately. If using an existing power supply, order cable assembly 32704-52.
$322.40 - $570.75USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Fluid Type Max Flow Rate (mL/min) Min Flow Rate (mL/min)
Availability Pricing
McMillan Flow 100-3 Ryton sensors for gases, 20 to 100 mL/min EW-32700-00 Mfr # 100-3
COMPARE
$323.50USD / Each
McMillan Flow 100-4 Ryton sensors for gases, 40 to 200 mL/min EW-32700-02 Mfr # 100-4
COMPARE
$323.50USD / Each
McMillan Flow 100-5 Ryton sensors for gases, 100 to 500 mL/min EW-32700-04 Mfr # 100-5
COMPARE
$323.50USD / Each
McMillan Flow 100-6 Economical Ryton PPS Flow Rate Sensor for Gases, 0.2 to 1 LPM (Air) EW-32700-06 Mfr # 100-6
COMPARE
$329.00USD / Each
McMillan Flow 100-7 Economical Ryton PPS Flow Rate Sensor for Gases, 0.4 to 2 LPM (Air) EW-32700-08 Mfr # 100-7
COMPARE
$324.70USD / Each
McMillan Flow 100-8 Economical Ryton PPS Flow Rate Sensor for Gases, 1 to 5 LPM (Air) EW-32700-10 Mfr # 100-8
COMPARE
$322.40USD / Each
McMillan Flow FS100-9 Economical Ryton PPS Flow Rate Sensor for Gases, 2 to 10 LPM (Air) EW-32700-12 Mfr # FS100-9
COMPARE
$419.30USD / Each
McMillan Flow 100-10 Economical Ryton PPS Flow Rate Sensor for Gases, 4 to 20 LPM (Air) EW-32700-14 Mfr # 100-10
COMPARE
$520.25USD / Each
McMillan Flow 100-11 Economical Ryton PPS Flow Rate Sensor for Gases, 10 to 50 LPM (Air) EW-32700-16 Mfr # 100-11
COMPARE
$520.25USD / Each
McMillan Flow 100-12 Economical Ryton PPS Flow Rate Sensor for Gases, 120 to 100 LPM (Air) EW-32700-18 Mfr # 100-12
COMPARE
$520.25USD / Each
McMillan Flow 100-13 Economical Ryton PPS Flow Rate Sensor for Gases, 40 to 200 LPM (Air) EW-32700-20 Mfr # 100-13
COMPARE
$536.25USD / Each
McMillan Flow 100-14 Economical Ryton PPS Flow Rate Sensor for Gases, 100 to 500 LPM (Air) EW-32700-22 Mfr # 100-14
COMPARE
$570.75USD / Each
McMillan Flow 101-3 Ryton sensors for liquids, 13 to 100 mL/min EW-32703-50 Mfr # 101-3
COMPARE
Water and other low-viscosity fluids (less than 10 centistokes)
$343.00USD / Each
McMillan Flow 101-5 Ryton sensors for liquids, 50 to 500 mL/min EW-32703-52 Mfr # 101-5
COMPARE
Water and other low-viscosity fluids (less than 10 centistokes)
$359.90USD / Each
McMillan Flow 101-6 Ryton sensors for liquids, 100 to 1000 mL/min EW-32703-54 Mfr # 101-6
COMPARE
Water and other low-viscosity fluids (less than 10 centistokes)
$359.90USD / Each
McMillan Flow 101-7 Economical Ryton PPS Flow Rate Sensor for Liquids, 0.2 to 2 LPM (Water) EW-32703-55 Mfr # 101-7
COMPARE
Water and other low-viscosity fluids (less than 10 centistokes)
$358.80USD / Each
McMillan Flow 101-8 Economical Ryton PPS Flow Rate Sensor for Liquids, 0.5 to 5 LPM (Water) EW-32703-56 Mfr # 101-8
COMPARE
Water and other low-viscosity fluids (less than 10 centistokes)
$422.30USD / Each
McMillan Flow 101-9 Economical Ryton PPS Flow Rate Sensor for Liquids, 1 to 10 LPM (Water) EW-32703-58 Mfr # 101-9
COMPARE
Water and other low-viscosity fluids (less than 10 centistokes)
$511.00USD / Each
Showing 1- 18 of 18