Economical Submersible Pumps, High-Flow Utility
Versatile – operate pumps in either a vertical or horizontal position
  • Compact and economical
  • Screened inlet helps prevent clogging
High-flow submersible utility pumps for use for water transfer with garden hose adpater or recirculation, water displays, air conditioners, and machine tool coolants. Removable inlet screen allows for water removal 1/8" from floor. Oil-filled for longer motor life. UL listed, CSA certified.
Note: Pumps must be fully submersed when operating and not for use with salt water or flammable solutions.
$270.20 - $293.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (m) Power (VAC)
Availability Pricing
COMPARE
20 8 115
$270.20USD / Each
COMPARE
20 8 230
$293.00USD / Each