Economical Submersible Pumps, Low-Flow Centrifugal
Versatile—operate pumps in either a vertical or horizontal position
  • Compact and economical
  • Screened inlet helps prevent clogging
Low-flow centrifugal submersible pumps for mild chemicals such as mild acids, alkalis, and hard water solutions compatible with pump materials. Use for water transfer or recirculation, water display, air conditioners, and machine tool coolants up to 100 SSU. Oil-filled for longer motor life.
Note: Pumps must be fully submersed when operating and not for use with salt water or flammable solutions.
$164.60 - $239.10USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Power (VAC)
Availability Pricing
COMPARE
3.5 10.2 115
$164.60USD / Each
COMPARE
5.4 12.3 115
$217.40USD / Each
COMPARE
5.4 12.3 230
$239.10USD / Each