Economical Submersible Pumps, Low- to Mid-Flow Centrifugal Submersible Pumps
Versatile – operate pumps in either a vertical or horizontal position
  • Compact and economical
  • Screened inlet helps prevent clogging
Low- to mid-flow centrifugal submersible pumps for water transfer or recirculation and water displays. Solution must be compatible with pump materials.
Note: Pumps must be fully submersed when operating and not for use with salt water or flammable solutions.
$129.40 - $328.20USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Max Head (ft) Wetted Materials
Availability Pricing
COMPARE
11 22.6 Aluminum, Nylon, Fluoroelastomer (FKM), Polypropylene, SS, PVC
$297.00USD / Each
COMPARE
2.8 4.6 Aluminum, polypropylene, nitrile, stainless steel, PVC
$129.40USD / Each
COMPARE
5 12.2 Polyester, Nylon, Fluoroelastomer (FKM), SS, PVC
$144.30USD / Each
COMPARE
5 11.8 Aluminum, Nylon, Nitrile, Polypropylene, SS, PVC
$159.90USD / Each
COMPARE
11.8 23.1 Aluminum, Nylon, Fluoroelastomer (FKM), SS, PVC
$328.20USD / Each