Ricca Chemical EDTA Titrant, 0.0575 Molar

Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$80.00 - $424.50USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Volume (mL) Description
Availability Pricing
Ricca Chemical Company 2710-32 EDTA Titrant, 0.0575 Molar (1 L) (Liquid) EW-86979-39 Mfr # 2710-32
COMPARE
Includes 6381-92-6 and 7732-18-5 1000 EDTA Titrant, 0.0575 Molar (1 L) (Liquid)
$80.00USD / Each
Ricca Chemical Company 2710-1 EDTA Titrant, 0.0575 Molar (1 gal) (Liquid) EW-86985-71 Mfr # 2710-1
COMPARE
Includes 6381-92-6 and 7732-18-5 3785 EDTA Titrant, 0.0575 Molar (1 gal) (Liquid)
$155.00USD / Each
Ricca Chemical Company 2710-2.5 EDTA Titrant, 0.0575 Molar (10 L) (Liquid) EW-86988-93 Mfr # 2710-2.5
COMPARE
Includes 6381-92-6 and 7732-18-5 10000 EDTA Titrant, 0.0575 Molar (10 L) (Liquid)
$300.25USD / Each
Ricca Chemical Company 2710-5 EDTA Titrant, 0.0575 Molar (20 L) (Liquid) EW-86993-32 Mfr # 2710-5
COMPARE
Includes 6381-92-6 and 7732-18-5 20000 EDTA Titrant, 0.0575 Molar (20 L) (Liquid)
$424.50USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat