Electro-Optic Point-Level Sensors
PSF wetted materials handles temperatures to 275°F!
  • Extremely compact- measures only 1.27" in length
  • Available in 1/4" NPT, 1/2"-20 SAE #5, and M12x1-8 mounting connections
Electro-optic sensors have no moving parts to wear out or clog in viscious liquids. Sensors have a ±1 mm repeatability and are suitable for high, medium, or low liquid detection. Solid state switching ensures dependable, long-life use. Typical applications include low lubricant, tank level, or reservior level warning. Order controller seperately below.
$84.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Depth (ft) Max Pressure (PSI) Min Depth (ft)
Availability Pricing
COMPARE
250
$84.00USD / Each
COMPARE
250
$84.00USD / Each